missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.8%, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 10037550 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle, packaged under nitrogen
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10037550 2.5 L Glass Bottle, packaged under nitrogen 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10037550 Supplier JT Baker Supplier No. 8402.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra gradient grade methanol for use in Liquid Chromatography (HPLC & UHPLC) and Spectrophotometry, packaged in 2.5L glass bottles.

BAKER ANALYZED™ HPLC grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Manufactured and tested to assure interference-free separations
  • Highly characterized reagents that have unmatched lot-to-lot consistency
  • Minimizes recalibration or instrument adjustments
  • Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
  • An Actual Lot Analysis is provided on the label

Specifications

Melting Point −98°C
Absorbance Max. 0.20 at 225nm, Max. 0.02 at 254nm, Max. 0.01 at 280nm, Max. 0.01 at 350nm
CAS 67-56-1
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 64.7°C
Quantity 2.5 L
Explosion Limit Lower: 5.5 vol %, Upper: 44 Vol %
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol
Packaging Glass Bottle, packaged under nitrogen
Physical Form Liquid
Residue after Evaporation Max. 2ppm
Molecular Formula CH4O
Titratable Acid Max. 0.0003meq/g
Titratable Base Max. 0.0001meq/g
MDL Number MFCD00004595
Water Max. 0.02%
Solubility Information Fully miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity ≥99.8%
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.