Learn More
MCT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20139-1-AP-20UL
 View more versions of this product
Product Code. 16899634
  • 1081.84 DKK / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Specifications
Specifications
MCT1
Polyclonal
Unconjugated
SLC16A1
MCT, MCT 1, MCT1, Monocarboxylate transporter 1, SLC16A1
Rabbit
Antigen Affinity Chromatography
RUO
6566
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53985
SLC16A1
MCT1 Fusion Protein Ag14098
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.