Learn More
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0
The 'MARCO' safety funnel can be safely closed and prevent danger of ignition, and is also available in conductive HDPE-EC material.
806.00 DKK - 1475.00 DKK
Product Code | Brand | Includes | Stopper Number | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Stopper Number | Height (Metric) | Price | Quantity & Availability | |||
16832140
|
SCAT™
318992 |
Funnel w/ Hinged Lid and Lance | GL45 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||
16609095
|
SCAT™
318952 |
Funnel w/ Hinged Lid, No Lance | GL45 | 150 mm |
885.00 DKK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||
16862140
|
SCAT™
318995 |
Funnel w/ Hinged Lid and Lance | S50 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||
16629095
|
SCAT™
318955 |
Funnel w/ Hinged Lid, No Lance | S50 | 150 mm |
815.00 DKK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||
16822130
|
SCAT™
318953 |
Funnel w/ Hinged Lid, No Lance | S51 | 150 mm |
815.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||
16842140
|
SCAT™
318993 |
Funnel w/ Hinged Lid and Lance | S51 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||
16822140
|
SCAT™
318991 |
Funnel w/ Hinged Lid and Lance | S55 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
|||
16699085
|
SCAT™
318951 |
Funnel w/ Hinged Lid, No Lance | S55 | 150 mm |
815.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||
16812140
|
SCAT™
318990 |
Funnel w/ Hinged Lid and Lance | S60/61 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||
16689085
|
SCAT™
318950 |
Funnel w/ Hinged Lid, No Lance | S60/61 | 150 mm |
815.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||
16852140
|
SCAT™
318994 |
Funnel w/ Hinged Lid and Lance | S65 | 370 mm |
910.00 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
16619095
|
SCAT™
318954 |
Funnel w/ Hinged Lid, No Lance | S65 | 150 mm |
815.00 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
17898723
|
SCAT™
318957 |
Funnel w/ Hinged Lid, No Lance | CPC/Justrite | 370 mm |
1475.00 DKK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||
17957260
|
SCAT™
318956 |
Funnel w/ Hinged Lid, No Lance | S70/71 | 150 mm |
806.00 DKK
Each |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||
17947250
|
SCAT™
318996 |
Funnel w/ Hinged Lid and Lance | S70/71 | 370 mm |
901.00 DKK
Each |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||
18626468
|
SCAT™
318958 |
Funnel w/ Hinged Lid, No Lance | 38-430 (Wheaton) | 150 mm |
807.00 DKK
1 sheet |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||
Description
- Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many chemicals
- Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
- Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents the escape of explosive or health-endangering gases
- Sturdy hinge guarantees a long service life of the closing mechanism
- Rotating screw cap makes it easier to screw the lid funnel on and off the canister
- Removable sieve for easy cleaning
- Non-conductive version of the MARCO V2.0 is in line with the SCAT color scheme due to its blue and white coloring
- Conductive versions of the MARCO V2.0 are in black and thus clearly differ in color from the non-conductive version
Specifications
300 mL | |
HDPE |