Learn More
3M™ Maintenance Sorbent Roll
A scrim on one side increases the strength and helps to eliminate linting. Available in convenient widths that are perfect for lining drawers, covering workbenches, or masking larger areas from mist and overspray.
Brand:  3M™ 7000051880
 View more versions of this product
Product Code. 17158600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Designed for use in and around the work area to catch leaks and drips
  • For use on most non-aggressive liquids
  • Very easy to use and to retrieve
  • Designed to cover large surfaces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
322.9 L
Workplace safety
45.7 x 0.96 m
45.7 m
960 mm
85.3 gal.
Sorbent Roll
M-B2002
150 ft.
38 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.