missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences MagStrep Strep-Tactin™ XT Beads
Tous les produits IBA Lifesciences Produits

Click to view available options
Capacité (métrique):
2 ml
10 ml
Conditionnement:
10mL
2mL
Description
- Can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.
- Beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-Tactin™ XT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP).
- MagStrep Strep-Tactin™ XT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.
- The beads can be easily and swiftly separated with a magnetic separator. This eliminates the need for centrifugation and reduces the risk of disturbing the pellet or loss of beads within the wash step to zero.
- The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.
Spécification
Spécification
Capacité (métrique) | 2 ml |
Formule | 5% (v/v) Suspension in PBS with 0.1% (w/v) Sodium Azide |
Type de support | 6% Agarose, Crosslinked |
Granulométrie | 20 to 40 μm |
Forme | Pâte de résine |
Cible | Twin-Strep-tag or Strep-tag II |
Type | Magnetic Agarose Bead |
Usage exclusivement réservé à la recherche
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu