missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Solution, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Boiling Point | >100°C |
| Melting Point | <0°C |
| CAS | 7786-30-3 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 L |
| Molecular Formula | Cl2Mg |
| Linear Formula | MgCl2 |
| MDL Number | MFCD00011106 |
| UN Number | NONH for all modes of transport |
| Show More |
Safety and Handling
Product Identifier
- Magnesium chloride solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction