Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
Includes:
100/Pk.
24/Pk.
Color:
Blue
Clear
Gray
Red
Yellow
Unit Size:
Pack of 100
Pack of 24
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
Specifications
| Color | Blue |
| Description | EFA: 7.2 cm2 |
| Material (Membrane) | PES |
| MWCO | 10 kDa |
| Sterility | Non-sterile |
| Includes | 24/Pk. |
| Volume (Metric) | 5 to 20 mL (Sample) |