missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane

Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.

Brand:  Cytiva MAP003C37

2313.25 DKK valid until 2025-12-31
Use promo code "24320" to get your promotional price.


 View more versions of this product

Product Code. 17144731

  • 2435.00 DKK / Pack of 24
Only null left
Estimated Shipment: 28-07-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gray
PES
Non-sterile
5 to 20 mL (Sample)
EFA: 7.2 cm2
3 kDa
24/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.