missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Bath
GREENER_CHOICE

Saves up to 80% against standard compressor units. Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Baths are supplied assembled as ready to use kits, complete with accessory hosing, clips and connectors as standard.

Brand:  Grant Instruments™ ECOCOOL 150R

33789.39 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.


 View more versions of this product

Product Code. 15419224

  • ONLINE EXCLUSIVE PRICE
    41280.00 DKK
    40266.80 DKK / Each
    Save 1013.20 DKK
    2% Off
View Available Promotions
Only null left
Estimated Shipment: 11-02-2026
Add to basket
This item is not returnable. View return policy

Description

Description

  • Eco mode optimises the cooling control needed, enabling rapid cool down when required.
  • True energy saving against competitor units that only switch the compressor on or off.
  • Intuitive user interface
  • No-spill valved front drain.
  • Built in Britain to the highest specifications, 5 day dispatch, 4 year industry leading warranty.
  • Modern, sleek, attractive design
  • Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing.
  • Single front switch for user convenience.
  • No side vents - locate to suit the user, not the unit.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 L
-25°C to +150°C
6 L
3
4 Years
154 mm
118 mm
120/230 V
400W at 20°C, 330W at 10°C, 200W at 0°C, 109W at -10°C, 60W at -10°C
Full Color QVGA TFT
ecocool™ 150
0.02°C
245 mm
640 mm
29 kg
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.