missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ 3 Fluorometer

Catalog No. 15387293
Change view
Click to view available options
Menge:
jeweils 1
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Menge unitSize
15387293 jeweils 1 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 15387293 Lieferant Invitrogen™ Lieferanten-Nr. Q33216

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Benötigt

500μL thin-walled PCR tubes

Accurately measures DNA, RNA, and protein using the highly sensitive fluorescence-based Qubit quantitation assays

The fluorescent dyes used in the Qubit assays emit signals only when bound to specific target molecules, even at low concentrations, thus minimizing the effects of contaminants, including degraded DNA or RNA. The integrated design of the instrument and assays results in quantitation that is far more sensitive than UV absorbance, making this system ideal for quantitation of precious samples (samples that are rare, difficult to purify, or expensive to either obtain or prepare) or samples for delicate applications (samples for downstream assays that are extremely sensitive to sample conditions).

Key Features

  • Powerful dual-core processor quickly and accurately quantifies DNA, RNA, and protein in less than 5 seconds per sample
  • Uses samples as little as 1μL up to 20μL
  • Stores up to 1000 sample results
  • Large, 5.7-inch, state-of-the-art color touch screen for easy workflow navigation
  • Ability to personalize Qubit fluorometer with the frequently run assays, add new assays, or even create custom assays with the MyQubit software and web tool
  • Small footprint saves bench space
  • Language options include English, French, Spanish, German, simplified Chinese and Japanese
  • Android-based operating system

More Accurate than UV Absorbance

  • More sensitive than UV absorbance, which measures anything absorbing at 260nm, including DNA, RNA, protein, free nucleotides, or excess salts
  • Additionally, UV spectrophotometry is often not sensitive enough to accurately measure low concentrations of DNA and RNA
  • The Qubit Fluorometer provides more accuarate measurements becuase the dyes in the Qubit assay kits fluoresce only when bound to the selected molecule—DNA, RNA, or protein—in a sample
User Friendly
  • Calculations and settings are automatically performed by the instrument
  • Qubit assays for use with the fluorometer are all performed using the same general protocol, which uses a simple mix-and-read format with incubation times of only two minutes
  • Advanced optics and data analysis algorithms
  • USB thumb drive and cable for data transfer to Excel™ sorftware and for software downloads

Applications:

Measurement of DNA, RNA, and protein using Qubit quantitation assays; Direct measurement of the fluorescence of samples; Evaluation of Ion Sphere™ Particle quality using the Ion Sphere Quality Control Kit prior to performing a sequencing run on the Ion PGM™ Sequencer

Zertifizierungen

Electromagnetic CE certification

TRUSTED_SUSTAINABILITY

Spezifikation

Probenvolumen 1 to 20 μL
Aufnahmevermögen 1 Assay Tube per Read
Excitation Wavelength 430 to 495 nm Blue LED, 600 to 645 nm Red LED
Zur Verwendung mit (Geräte) Qubit Fluorometer
Produktlinie Qubit
Typ Fluorometer
Displaytyp 5.7 in. Capacitive Touchscreen
Zur Verwendung mit (Anwendung) DNA-Quantifizierung, RNA-Quantifizierung, Proteinquantifizierung
Hauptspeicher 1000 Samples
Versandbedingung Raumtemperatur
Abmessungen (L x B x H) 10 x 5.4 x 2.2 in. (25 x 13.6 x 5.5 cm)
Gewicht (metrisch) 743.0 g
Spannung 100/240 VAC
Menge jeweils 1
Mehr anzeigen Weniger anzeigen

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.