missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MP Biomedicals™

Product Code. 11319009 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319009 1 kg 1kg
11388999 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319009 Supplier MP Biomedicals Supplier No. 0210214791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Molecular biology, Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8030-76-0
Molecular Formula C11H22NO8P
Synonym L-a-Phosphatidylcholine

Specifications

Color Yellow
Quantity 1 kg
Formula Weight 327.27
Physical Form Solid
Chemical Name or Material Lecithin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.