missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Staphylococcus aureus
Shop All Novacyt Group ProductsDescription
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | S. aureus |
| Type | Latex Agglutination Test |
| Quantity | 100 Tests |
| Test Time | 2 min. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?