missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Serum Bottle Aluminum Seals, Large

Product Code. 12373680 Shop All Labconco Corporation Products
Change view
Click to view available options
Thread Size (Metric):
57 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
12373680 57 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12373680 Supplier Labconco™ Supplier No. 7577110
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accessory for freeze dry glassware

  • Fits 5mL, 10mL, 20mL, 30mL, 50mL, 100mL and 125mL serum bottle

Specifications

Color Silver
For Use With (Application) Fits 5mL, 10mL, 20mL, 30mL, 50mL, 100mL and 125mL Serum bottles
Material Aluminum
Sterility Non-sterile
Weight (Metric) 0.5kg
Weight (English) 1 lb.
Thread Size (Metric) 57 mm
Neck Finish 57 mm
Disposable Single-use
Product Type Cap
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.