missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Fast-Freeze™ Flask Adapter

Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.

Brand:  Labconco™ 7547200

325.22 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10196103

  • 351.00 DKK / Each
Only null left
Estimated Shipment: 29-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

For connecting Fast-Freeze™ flasks to freeze-dryer valves

  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications

Specifications

Flask Adapter
Stainless Steel
For connecting fast freeze™ flasks to freeze-dryer valves
0.75 in.
0.75 in.
3.25 in.
0.5 lb.
40, 80 mL
Straight
Fast freeze™ flask
1.9 cm
1.905 cm
8.26 cm
0.2 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.