Learn More
Berkshire Lab-Tips™ Pointed Head Swab
The LTO70P Swab with pointed head is an excellent swab for precision cleaning applications.
Brand:  Berkshire LT070P20
3478.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17887290
  • 4125.00 DKK / Pack of 2000
Only null left
Estimated Shipment: 05-12-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Cleanroom laundered polyurethane foam over rigid tip
  • 100 ppi open cell foam
  • Polypropylene handle with thermally bonded head
  • Laundered and packaged in Class 10 {ISO Class 4} cleanroom
  • Low particles, fibers, ions and extractables
  • Good chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Free from Silicone, amides and DOP
  • Ridged surface for improved cleaning ability
Specifications
Specifications
2.75 in.
Not Autoclavable
Plastic
Foam
Cleanroom Swab
Thermal
Pointed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.