Learn More
Thermo Scientific™ KingFisher™ Pure DNA Blood Kit
Achieve rapid, reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate with this unique nucleic acid purification workflow.
Specificatie
| Monstertype | Fresh or frozen whole blood treated with EDTA or citrate |
|---|---|
| Geschikte downstreamanalyse | PCR, Real-time PCR, Enzymatic Reactions |
| Elutievolume (metrisch) | 200µL sample: 100µL (Duo) or 150µL (Flex). 1mL sample: 500µL. Recommended but adjustable |
Beschrijving
Achieve rapid, reproducible purification of high-quality DNA. The Thermo Scientific™ KingFisher™ Pure DNA Blood Kit, together with a KingFisher magnetic particle processor, allows rapid and reproducible purification of high-quality DNA from fresh or frozen whole blood treated with EDTA or citrate. Thermo Scientific KingFisher Kits complete the unique nucleic acid purification workflow providing an optimized high throughput method for extreme flexibility.
Features- Produces isolated DNA that is free of inhibitors and ready for use in many downstream applications
- Easy-to-use protocols for safe sample-handling, as well as minimal user input, maximizing walkaway time.
- Excellent performance and consistency of results optimized to produce high yields of quality DNA
- Large volume sample input possible up to 3mL of blood per sample
- Flexible throughput from one sample to up to 96 samples per run
- Minimized hands-on time increases efficiency
- Compatible with most common downstream analysis and applications
Recommended for:
Genomics, target identification, veterinary assays, biomarker discovery, quality control
Specificatie
| Fresh or frozen whole blood treated with EDTA or citrate | |
| PCR, Real-time PCR, Enzymatic Reactions | |
| 200µL sample: 100µL (Duo) or 150µL (Flex). 1mL sample: 500µL. Recommended but adjustable |
Veiligheid en behandeling
- MIXTURE LIST-Bevat: Sodium perchlorate,Triton X-100, Guanidine hydrochloride, Proteinase K
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.