Learn More
Kinematica Polytron™ Standard Dispersing Aggregates
Kinematica Polytron Standard Dispersing Aggregates, Bottom Type: Saw tooth, Diameter: 12 mm, For Use With: PT 1200 E, PT 1300 D and PT 2500 E, Length: 157 mm, Stainless Steel, Sample Volume: 2 to 250 mL
Brand:  Kinematica 11030042
Product Code. 10364972
  • ONLINE EXCLUSIVE PRICE
    1965.00 €
    1913.00 € / Each
    Save 52.00 €
    2% Off
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Kinematica Polytron Standard Dispersing Aggregates, Bottom Type: Saw tooth, Diameter: 12 mm, For Use With: PT 1200 E, PT 1300 D and PT 2500 E, Length: 157 mm, Stainless Steel, Sample Volume: 2 to 250 mL
Specifications
Specifications
Standard Dispersing Aggregate
2 to 250 mL
157 mm
Sawtooth
250 mL
12 mm
Stainless Steel
PT 1200 E, PT 1300 D, PT 2500 E
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.