Learn More
Kinematica™ Polytron™ PT1200E Handheld Homogenizer
This ergonomic handheld homogenizer is lightweight and therefore ideal for rapid turn-around of smaller sample volumes. For sample volumes 0.05 ml to 250 ml.
Brand: Kinematica™ 11010025
Description
This ergonomic handheld homogenizer is lightweight and therefore ideal for rapid turn-around of smaller sample volumes. For sample volumes 0.05 ml to 250 ml.
- Easy operation with an on/off switch and speed control
- Intelligent variable speed control for long equipment life
- Low noise operation
- Height adjustable stand (available as an option)
- Easy clean-up
- Smallest size of particles/droplets because precise analysis requires homogeneous samples
- Efficient sample preparation because fast dispersion saves time
- Excellent for general homogenizing applications, dispersions, suspensions, emulsions, cell breakdowns and more
- Universal motor with carbon brushes
- 80% relative max. humidity during storage/operation
- IP protection class IP 20
Specifications
| Drive Unit | |
| 100 W | |
| 0.05 to 250 mL | |
| 55 mm | |
| PT 1200E | |
| Drive: 440 g, power supply: 310 g |
| 0 to 25,000 rpm | |
| 250 mL | |
| 220 mm | |
| Ideal For Pulverizing Plant And Animal Tissue, Grain And Seed, Soil Extraction, DNA/RNA Extraction, Karl Fischer Sample Prep, Toxicology And Food Science | |
| 0°C to 40°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.