missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Science Precision Wipes
GREENER_CHOICE

Low-lint, high absorbency and chemically inert wipes for delicate tasks and instrumentation in a range of applications. The 1-ply white-coloured wipes are provided in a convenient pop-up box and are anti-static.

Brand:  Kimberly-Clark™ 7551

31.61 EUR valid until 2025-09-30
Use promo code "24298" to get your promotional price.


Product Code. 12660543

  • € 57.00 / Pack of 198
Spotlight
Use promo code 24158 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket

Description

Description

  • For delicate equipment, surfaces, optics and materials, our low-lint and high absorbency white wipes have you covered
  • Light duty, white-colored wipes made from 100% pure fiber
  • Provided in a hygienic, user-friendly pop-up box
  • Low-lint absorbency for delicate applications and surfaces
  • Suitable for use in ISO Class 4 or higher environments
  • Suitable for applications in healthcare, manufacturing, optics and electronics
  • Chemically inert and anti-static
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Critical Task Wipe
28.5 cm
White
1-ply
31 cm
Cellulose
Light Duty, Precision Cleaning of Delicate Glassware, Instruments and Surface/Parts
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.