Learn More
Kimtech Science™ Wipe Delicate Task White
GREENER_CHOICE
Wipe liquids with multi-ply absorbency and low lint content for intricate dusting. Kimberly Clark™ Kimtech Scientific™ Delicate Task Wipers are ideal for cleaning delicate glassware, instruments, and surface/parts.
Brand:  Kimberly Clark 7557
88.46 EUR valid until 2025-09-30
Use promo code "24298" to get your promotional price.
Product Code. 17701868
  • € 154.99 / Pack of 2400
Spotlight
Use promo code 24158 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
A range of high-performance wipers, suitable for ISO Class 4 or higher laboratory environments.
Ideal for: cleaning of delicate glassware, instruments and surface/parts; wiping liquids, with multi-ply absorbency and low lint content for intricate dusting.
Available as: 100 or 200 format, white paper wipers, dispensed from a POP-UP Box flexible system, giving controlled access to hygienically protected wipers when you’re cleaning or wiping on the move.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wipe
21 cm
White
Suitable for ISO Class 4 or higher laboratory environments.
2-ply
20 cm
Paper
Wiping liquids or cleaning delicate glass, instruments, parts, and surfaces.
100 Sheets x 24 Dispenser Boxes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.