missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, purum p.a, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.50 L
5 L
Specifications
Specifications
Color | Colorless |
Density | 0.8g/mL at 25°C |
Boiling Point | 175°C to 3257deg;C |
Packaging | Glass bottle |
Quantity | 1 L |
UN Number | UN1223 |
Vapor Density | 4.5 (vs air) |
Physical Form | Liquid |
Chemical Name or Material | Kerosene |
Safety and Handling




Product Identifier
- Kerosene
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Kerosene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction