missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 70%, Honeywell™

Artikelnummer. 15685840 Shop alle Honeywell Research Chemicals produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 L
200 L
Packaging:
Glass Bottle
Poly Drum
Pakningsstørrelse:
1 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
15685840 1 L Glass Bottle 1 L
15695840 200 L Poly Drum 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15685840 Leverandør Honeywell Research Chemicals Leverandørnr. 5639351L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

70% in H2O

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Tekniske data

Density 0.858g/mL at 25°C
Quantity 1 L
Packaging Glass Bottle
UN Number UN1219
Chemical Name or Material Isopropanol
compliance-icons compliance-icons
Produktidentifikator
  • Isopropanol
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
Faresætning
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contains : Propan-2-ol
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.