missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Safety Sign Roll without Text - Warning: Biological Hazard
Safety sign for hazard identification.
Brand: Brady™ 827648
989.44 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
PIC W009- Tri 050-PE-ROLL/1 | |
Laminated Polyester | |
Triangular | |
Black on Yellow |
Gloss | |
Permanent Acrylic | |
43 x 50 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction