Learn More
Invitrogen™ Lipofectamine™ 2000 CD Transfection Reagent
Description
These reagents allow highly efficient transfection of broad range of cell types, including adherent, suspension, insect, and endothelial lines, as well as primary cultures. All transfection reagents include streamlined protocols for maximum convenience and ease of use in many applications, including high-throughput applications.
- Highest transfection efficiency in many cell types and formats
- Animal origin-free formulation ensures that mammalian cell culture and bioproduction processes are free of animal-derived materials
- DNA-Lipofectamine 2000 CD complexes can be added directly to cells in culture medium
- It is not necessary to remove complexes or change/add medium after transfection, but complexes may be removed after 4–6 hours
PCR and Real-Time PCR, Reverse Transcription, cDNA Synthesis Direct from Cells
Order Info
Shipping Condition: Qualified for Ambient or Wet Ice
Specifications
Specifications
| Classification | Animal Origin-Free, Chemically Defined |
| Content And Storage | Each reagent should be stored at 2–8°C upon receipt. |
| Cell Type | Established Cell Lines |
| Sample Type | Plasmid DNA |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | Lipofectamine |
| Product Type | Transfection Reagent |
| Quantity | 1 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.