Learn More
Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
€ 117.00 - € 196.00
Product Code | Brand | Tube Size | No. of Places | Depth (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Size | No. of Places | Depth (Metric) | Price | Quantity & Availability | |||
11490908
|
Grant Instruments™
QB-0 |
None | Solid Block | None |
€ 120.00 € 117.00 / Each Save € 3.00 2% Off |
In Stock
Log in to see stock. ![]() |
|||
10211643
|
Grant Instruments™
QB-50 |
50mL | 4 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||
10781434
|
Grant Instruments™
QB-24 |
24mm | 5 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
12664915
|
Grant Instruments™
QB-17H |
17mm | 10 | 75 mm hole depth |
€ 201.00 € 196.00 / Each Save € 5.00 2% Off |
In Stock
Log in to see stock. |
|||
11420918
|
Grant Instruments™
QB-13 |
13mm | 12 | 50 mm hole depth |
€ 162.74 € 158.00 / Each Save € 4.74 2% Off |
In Stock
Log in to see stock. |
|||
11430918
|
Grant Instruments™
QB-16 |
16mm | 12 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
15389625
|
Grant Instruments™
QB-E5 |
16.7 mm | 12 | 53.5 mm hole depth | |||||
11440918
|
Grant Instruments™
QB-18 |
18mm | 12 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11480918
|
Grant Instruments™
QB-E0 |
0.5mL | 24 | 30 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11400918
|
Grant Instruments™
QB-10 |
10mm | 24 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11490918
|
Grant Instruments™
QB-E1 |
1.5mL | 24 | 35 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11400928
|
Grant Instruments™
QB-E2 |
2.0mL | 24 | 35 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11410918
|
Grant Instruments™
QB-12 |
12mm | 24 | 50 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
11470918
|
Grant Instruments™
QB-H |
0.2mL | 56 | 14 mm hole depth |
€ 162.00 € 158.00 / Each Save € 4.00 2% Off |
In Stock
Log in to see stock. |
|||
Description
- For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
- Accommodates a variety of test tube sizes
- Available in plain block format without holes