missing translation for 'onlineSavingsMsg'
Learn More

Instrument Calibration Standard 1 For ICP/MS, SPEX CertiPrep™

Product Code. 15618245 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15618245 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15618245 Supplier SPEX Certiprep Supplier No. CLCAL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag20μg/mL
Al20μg/mL
As20μg/mL
Ba20μg/mL
Be20μg/mL
Cd20μg/mL
Co20μg/mL
Cr20μg/mL
Cu20μg/mL
Mn20μg/mL
Mo20μg/mL
Ni20μg/mL
Pb20μg/mL
Sb20μg/mL
Se20μg/mL
Th20μg/mL
Tl20μg/mL
U20μg/mL
V20μg/mL
Zn20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7697-37-2
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula HNO3
MDL Number MFCD00011349
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Type ICP/ICP-MS Standard
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Solvent or Matrix 5% HNO3/tr Tart. Acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
Quantity 125 mL
ChEBI CHEBI:48107
Grade ICP/ICP-MS
Concentration or Composition (by Analyte or Components) Each component at 20μg/mL in 2% HNO3/tr. Tartaric Acid:, Aluminum, Antimony, Arsenic, Barium, Beryllium, Cadmium, Chromium, Cobalt, Copper, Lead, Manganese, Molybdenum, Nickel, Selenium, Silver, Thallium, Thorium, Uranium, Vanadium, Zinc
Chemical Name or Material Instrument Calibration Standard 1
Show More Show Less
compliance-icons
Product Identifier
  • instrument calibration standard 1
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitic acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.