missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert 1/4 for Laboratory Dishwasher

Insert E 103/1 for the optimum loading of test tubes up to 12 x 75mm.

Brand:  Miele Professional™ 12667340

Product Code. 10282641

  • ONLINE EXCLUSIVE PRICE
    € 334.00
    € 281.00 / Each
    Save € 53.00
    15% Off
Only null left
Estimated Shipment: 27-11-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for up to 200 test tubes
  • divided into 6 surfaces
  • the mesh width is 8 x 8mm
  • including lid A 13
  • For use with PG8583, PG8593
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mesh basket
Upper or lower baskets
102 mm
1 Each
A 13
320 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.