missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Routine pH Electrodes

A combination pH electrode with an integrated temperature sensor and chemically resistant HA glass body. It is designed for general lab applications and ideal for use at high pH values (IP67 rated).

Brand:  METTLER TOLEDO™ 51343056

315.32 EUR valid until 2025-10-26
Use promo code "24136" to get your promotional price.


 View more versions of this product

Product Code. 10212283

  • ONLINE EXCLUSIVE PRICE
    € 343.00
    € 334.00 / Each
    Save € 9.00
    2% Off
View Available Promotions
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Drinking water
  • Highly saline solutions or seawater
  • Microbiological samples
  • Corrosive acids and bases
  • General Lab
  • Chemical
  • High pH values
  • TRIS compatible

Accessories & Care

  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Tip: Open fill port while measuring, and also to refill electrolyte solution
  • Ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Order Info

Cable not included (sold separately).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multi-Pin
Glass
0°C to 100°C
3 mol/L KCl
Multi-Pin to BNC (30281896)
ARGENTHAL
InLab™ Routine Pt1000
<20 sec.
32°F to 212°F
4.72 in.
0.47 in.
Ceramic
0 to 14
Combination Electrode
Accurate measurement in a wide range of sample types
Temperature Probe (Pt1000)
HA Glass
PT1000 ATC
3 mol/L KCl
0°C to 100°C
120 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.