missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva NAP™ Columns, NAP-5
Shop All Cytiva Products
Click to view available options
Quantity:
20 Pack
50 Pack
Unit Size:
Pack of 20
Pack of 50
Description
- Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
- Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
- Useful for small-scale purification, desalting, and buffer exchange

Specifications
Specifications
For Use With (Application) | Rapid and efficient purification of DNA and Oligonucleotides |
Includes | Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution |
Product Type | NAP-5 |
Quantity | 50 Pack |
Safety and Handling

Signal Word
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction