missing translation for 'onlineSavingsMsg'
Learn More

R 270 Boss Head Clamp

Secure joints with this clamp. IKA™ R 270 Boss Head Clamp features a sturdy, cast aluminum construction and cross sleeve design. Ideal for a variety of applications.

Brand:  IKA 0002657800

135.92 EUR valid until 2025-10-31
Use promo code "24158" to get your promotional price.


144.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11461178

  • ONLINE EXCLUSIVE PRICE
    € 149.76
    € 144.00 / Each
    Save € 5.76
    3% Off
View Available Promotions
Only null left
In Stock
Edge
Add to basket

Description

Description

  • Clamping range, stand: 25 to 36mm
  • Clamping range, extension arm: 5 to 21mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Boss Head Clamp
Cast Aluminum
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.