Learn More
IBA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10904-1-AP-150UL
 View more versions of this product
Product Code. 16897593
  • 3058.99 DKK / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.
Specifications
Specifications
IBA1
Polyclonal
Unconjugated
AIF1
AIF 1, AIF1, G1, IBA1, IRT 1, Protein G1
Rabbit
Antigen Affinity Chromatography
RUO
11629, 199
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70200, P55008
AIF1
IBA1 Fusion Protein Ag1363
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.