missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-819 Nylon/Spandex ESD Gloves with FORTIX™ Nitrile Foam Coating
GREENER_CHOICE

Safety gloves with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. Ultra-thin FORTIX™ coating enhances grip, while also maintaining bare hand sensitivity for tasks that require dexterity and precision.

908.00 DKK

Specifications

Certifications/Compliance (Europe) EN 16350:2014, EN 420:2003 + A1:2009
Coated Area Palm
Applications Industrial
Coating Material Nitrile Foam
Color Blue
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17270157
GREENER_CHOICE
View Documents
Ansell
11819060
6
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 17-12-2025
Add to basket
 
17280157
GREENER_CHOICE
View Documents
Ansell
11819070
7
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 02-10-2025
Add to basket
 
17290157
GREENER_CHOICE
View Documents
Ansell
11819080
8
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 02-10-2025
Add to basket
 
17200167
GREENER_CHOICE
View Documents
Ansell
11819090
9
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 09-10-2025
Add to basket
 
17210167
GREENER_CHOICE
View Documents
Ansell
11819100
10
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 16-09-2025
Add to basket
 
17220167
GREENER_CHOICE
View Documents
Ansell
11819110
11
908.00 DKK
12 pairs
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 17-12-2025
Add to basket
 
Description

Description

  • ESD functionality certified to EN 16350:2014
  • Can be used in explosion hazard zones as part of a grounding chain
  • Seamless, 18 gauge nylon and spandex liner helps maintain a bare-handed feel for tasks that require dexterity and precision
  • FORTIX™ nitrile foam coating enhances grip and provides long lasting protection
  • ZONZ™ Comfort Fit Technology ensures gloves are ultra breathable
  • Proprietary washing process helps to deliver cleaner, more comfortable gloves
  • Touchscreen functionality allows users to work with multiple touchscreen devices without glove removal
  • Dermatologically approved by Dermatest™
  • Certified by OEKO-TEX™
Specifications

Specifications

EN 16350:2014, EN 420:2003 + A1:2009
Industrial
Blue
Specific
Nylon, Spandex
Non-sterile
Aerospace, Automotive, Logistics, and Machinery/Equipment
Coated
18
Palm
Nitrile Foam
Reusable
Yes
ESD Gloves
3121A
Knit
HyFlex™ 11-819
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.