missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
10 L
2.5 L
5 L
Description
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | HDPE plastic bottle |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| IUPAC Name | chlorane |
| Formula Weight | 36.46g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction