missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™

Product Code. 15634420 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634420 1 L 1 L
15664420 500 mL 500mL
15644420 2.5 L 2.5 L
15684420 6 x 500 mL Pack of 6
15674420 6 x 1 L Pack of 6
15654420 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15634420 Supplier Honeywell-Hydranal Supplier No. 348011L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Titrant 5
Density 0.85g/mL
Quantity 1 L
Flash Point 11°C (51.8°F)
UN Number UN1230
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.,Toxic in contact with skin.
  • H370-Causes damage to organs.,Causes damage to organs.
  • H331-Toxic if inhaled.,Toxic if inhaled.
  • H373-May cause damage to organs.,May cause damage to organs.
Precautionary Statement
  • P373-DO NOT fight fire when fire reaches explosives.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, iodine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.