missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™

Brand:  Honeywell Fluka 34800-4X2.5L


 View more versions of this product

Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15694410

  • 5937.66 DKK / Pack of 4
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Solvent
11°C (51.8°F)
MFCD00147197
0.89g/mL
4 x 2.5 L
UN3286
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-Solvent

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.