missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™
Shop All Honeywell Fluka ProductsSpecifications
Specifications
| Chemical Name or Material | HYDRANAL™-Solvent |
| CAS | 67-56-1, 288-32-4, 7446-09-5 |
| Density | 0.89g/mL |
| Quantity | 1 L |
| Flash Point | 11°C (51.8°F) |
| MDL Number | MFCD00147197 |
| UN Number | UN3286 |
Safety and Handling
Product Identifier
- HYDRANAL-Solvent
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.,May damage the unborn child.,May damage the unborn child.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.,Causes damage to organs.,Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: methanol, imidazole,sulphur dioxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?