missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™

Product Code. 15654500 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Kit
10 x 5 mL Ampules
6 x 25 mL
Unit Size:
50mL
Pack of 6
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15654500

Brand: Honeywell-Hydranal 3484050ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Specifications

Chemical Name or Material HYDRANAL™-Coulomat CG
CAS 134227-25-1, 67-56-1, 14426-21-2, 7446-09-5
Quantity 10 x 5 mL Ampules
Flash Point 14°C (57.2°F)
UN Number UN1230
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat CG
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, diethanolamine hydrochloride,Ethanol, 2,2'-iminobis-, hydriodide, sulphur dioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.