missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AK, Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka

Brand:  Honeywell-Hydranal 34820-500ML


 View more versions of this product

Product Code. 15684480

  • 340.46 EUR / 500mL
Spotlight
Use promo code 24156 at checkout
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - Coulomat AK
1.200 g/cm3
500 mL
≥30.0000%, ≥20.0000%, ≥10.0000%, ≥5.0000%, ≥1.0000%, ≥1.0000%
60°C
UN1992
Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm
32°C
109-86-4, 67-66-3, 75-89-8, 101023-55-6, 68007-08-9, 7446-09-5
<50.0000%, <30.0000%, <20.0000%, <10.0000%, <5.0000%, <5.0000%
Glass Bottle
Karl Fischer
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 2-methoxyethanol; ethylene glycol monomethyl ether, trichloromethane; chloroform, 2,2,2-Trifluoroeth

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.