missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka

Product Code. 15614390 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15614390 500 mL 500mL
15624390 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 15614390 Supplier Honeywell-Hydranal Supplier No. 34739500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat AG-Oven
Name Note Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm
Boiling Point 64°C
CAS Min % ≥5.0000%, ≥20.0000%
CAS Max % <20.0000%, <10.0000%, <10.0000%, <30.0000%
CAS 111-42-2, 7446-09-5, 57-55-6, 288-32-4
Density 0.980 g/cm3
Quantity 500 mL
Flash Point 13°C
Packaging Glass Bottle
UN Number UN1230
Grade Karl Fischer
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat AG-Oven
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, Propane-1,2-diol, 2,2'-iminodiethanol; diethanolamine, imidazole, sulphur dioxide, 1H-Imid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.