missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat AD, Reagent for coulometric KF titration (anolyte solution), for cells without diaphragm, Honeywell Fluka™
Shop All Honeywell Fluka ProductsSpecifications
Specifications
| Chemical Name or Material | HYDRANAL™-Coulomat AD |
| CAS | 7446-09-5, 68007-08-9, 288-32-4 |
| Quantity | 500 mL |
| Flash Point | 11°C (51.8°F) |
| UN Number | UN1230 |
Safety and Handling
Product Identifier
- HYDRANAL-Coulomat AD
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: methanol, 2,2'-iminodiethanol; diethanolamine, sulphur dioxide, imidazole, 1H-Imidazole monohydriodi
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?