missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™

Brand:  Honeywell-Hydranal 34816-2.5L


 View more versions of this product

Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15694470

  • 2868.22 DKK / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Composite 5 K
90°C (194°F)
111-90-0, 7553-56-2, 288-32-4, 693-98-1, 7446-09-5, 68007-08-9
1.15g/mL
2.5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL Composite 5 K

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : iodine, imidazole, sulphur dioxide, 1H-Imidazole monohydriodide, 2-Methylimidazole

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.