missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Composite 1 |
| CAS | 288-32-4, 7446-09-5, 111-90-0 |
| Density | 1.11g/mL |
| Quantity | 1 L |
| Flash Point | 90°C (194°F) |
Safety and Handling
Product Identifier
- HYDRANAL-Composite 1
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, imidazole, 2-Methylimidazole, sulphur dioxide, iodine, 1H-Imidazole monoh
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction