missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Buffer cell culture with PBS in solution.

Brand:  Cytiva SH30264.02

 View more versions of this product

Product Code. 10540853

  • 32.40 € / 1000mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Manufactured under cGMP and ISO 9001 certified facilities
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) With Calcium, Magnesium, without Phenol Red
Liquid
Sterile
Calcium, Magnesium
≤0.025EU/mL
276±10mOsm/kg H2O
1000 mL
15° to 30°C
1 X
Dulbecco's Phosphate Buffered Saline
0.1 μm Sterile-filtered
No Phenol Red
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
7 to 7.6
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.