Learn More
HTR6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23561-1-AP-150UL
 View more versions of this product
Product Code. 16872385
  • € 410.00 / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.
Specifications
Specifications
HTR6
Polyclonal
Unconjugated
HTR6
5 HT 6, 5 HT6, 5 HT6R, 5 hydroxytryptamine receptor 6, HTR6, Serotonin receptor 6
Rabbit
Antigen Affinity Chromatography
RUO
3362
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50406
HTR6
HTR6 Fusion Protein Ag15396
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.