Learn More
SCAT HPLC Starter Kits - Supply, V2.0
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
Brand:  SCAT 399203
 View more versions of this product
Product Code. 18656488
  • ONLINE EXCLUSIVE PRICE
    3105.00 DKK
    2439.73 DKK / 1 set
    Save 665.27 DKK
    21% Off
Only null left
Estimated Shipment: 13-11-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Original brand quality from the premium manufacturer SCAT
  • Initial removal equipment that can be used with all common HPLC system
  • Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
  • All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
Specifications
HPLC Starter Kit 4: 4 x Safety Cap IV, GL45, 4 x air valve, 4 x change indicator, 16 x PFA fitting 1.6 mm OD, green, 16 x PFA fitting 2.3 mm OD, violet, 16 x PFA fitting 3.2 mm OD, blue, 12 x PFA blind plugs, 4 x labelling field
4 Pk.
Solvent Waste Kit
PTFE
GL45
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.