Learn More
SCAT HPLC Starter Kits - Supply, V2.0
SCAT Safety Caps are the standard for safely supplying your HPLC with solvents. They block harmful vapours and protect your HPLC system from contamination.HPLC starter kit equipped with SafetyCaps, blanking plugs, fittings and vent valves.
Brand: SCAT 399201
2162.29 DKK valid until 2025-08-31
Use promo code "24215" to get your promotional price.
Description
- Original brand quality from the premium manufacturer SCAT
- Initial removal equipment that can be used with all common HPLC system
- Starter kit consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems
- All set components can be reordered separately
starter kit, hplc system, safetycaps, blind plugs, fittings, air valve
Specifications
Supply of HPLC Systems | |
4 x SafetyCaps II (307909), 4 x blind plugs, 8 x fittings 1.6/2.3/3.2 mm, 4 x air valves (317010) | |
HPLC Starter Kit 2 |
Any HPLC System | |
GL45 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.