Learn More
Stuart™ Hotplate Accessories
Product Code. 15135934
Change view
Click to view available options
Size:
Small
5 mL
10 mL
25 mL
Type:
Block Base
Block Insert
Clamp, Heater Block
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type unitSize
15135934 Small Clamp, Heater Block Each
15125914 5 mL Block Insert Each
15105924 10 mL Block Insert Each
15135924 25 mL Block Insert Each
15105914 Small Block Base Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15135934 Supplier Stuart™ Supplier No. 0480572
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Hotplate Accessories for use with Hotplates/Stirrers that have a 6 x 6 in. top plate.
  • This heating block system with interchangeable inserts can be used with various Stuart Hotplates and Hotplate Stirrers
  • Designed to heat round-bottom flasks
  • Small and medium sized base plates offered; each can accommodate up to three block inserts each
  • A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL
  • Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions
  • Hotplate accessories offer equal or better heating performance than silicon oil
  • The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware
  • Each base plate features a hole to house a temperature controller probe
Specifications
For Use With (Equipment) Hot Plates (6 x 6 in.)
Size Small
Type Clamp, Heater Block