Learn More
Kinematica™ Polytron™ PT 10-35 GT Homogenzier Drive Unit
Compact,economical homogenizer with 1200 W motor for processing larger samples. Versatile design allows for samples as small as .1 ml and up to 4500 ml with appropriate dispersing aggregate. Over 25 aggregates are available for almost any dispersing or homogenizing application.
Brand: Kinematica™ 11010104
Description
- Application volumes 0.1mL to 10L
- 1200W motor provides sufficient power and high speed necessary for efficient particle size reduction
- Digital motor speed control from 500 to 30,000 rpm
- Aggregate tip speeds up to 32 m/s produce the finest particles possible
- Soft start prevents splashing of homogenate from sample vessel
Specifications
| Homogenizer Drive Unit | |
| 230 V | |
| 0.1 mL to 10 L | |
| Difficult-to-process Samples |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.