missing translation for 'onlineSavingsMsg'
Learn More

Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™

Product Code. 15694560
Change view
Click to view available options
Quantity:
1 L
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15694560 2.5 L 2.5 L
15684560 1 L 1 L
15604570 2.5 L-PC 2.5 L
15624570 6 x 1 L Pack of 6
15614570 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15694560 Supplier Honeywell-Chromasolv Supplier No. 348592.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CHROMASOLV™, for HPLC,≥97.0% (GC)

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Spécification

Melting Point -95°C
Density 0.659g/mL at 25°C
Boiling Point 69°C
Quantity 2.5 L
Refractive Index n20/D 1.375
Linear Formula CH3(CH2)4CH3
Vapor Pressure 256mmHg (37.7°C)
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥97.0% (GC)
Grade CHROMASOLV
Chemical Name or Material Hexane, for HPLC
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Hexane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contains : n-hexane
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.