Learn More
Thermo Scientific™ Heratherm™ IMP400 Refrigerated Incubator, 381 L, Stainless Steel
Temperature Range: 5°C to 70°C
Brand: Thermo Scientific™ 51031565
9504.28 EUR valid until 2025-10-31
Use promo code "24158" to get your promotional price.
Description
Achieve untapped potential in incubation with the temperature precision you need for applications that demand it.. With a temperature range of 5-70°C, Thermo Scientific Heratherm Refrigerated Incubators are equipped with Peltier technology to reach and maintain the precise ambient temperatures you need for reliable results. With no hazardous refrigerants, these units use less energy than the conventional product, making it the ideal choice for energy-conscious labs
- Temperature range +5 to +70°C
- Available in 2 sizes: benchtopmodel 178L / 6.3 cuft and a floormodel with casters 381L / 13.5 cu.ft.
- Great temperature uniformity and stability to keep the samples in a safe environment –> uniformity as low as +/-0.3°C (20°C), stability as low as +/-0.1°C
- Energy consumption as low as 80Wh/h (bench top) / 150Wh/h(floor) at 20°C - Note that comparable compressor cooled units consume up to 6 times this amount*.
- CFC free / HFC free polyurethane insulation foam for great temperature performance and condensate prevention
- Heated door with intelligent energy control to ensure no condensation – even at low temperatures
- Sophisticated timer function with weekly, daily or real time mode
- Intuitive programmability function for defined temperature ramping, soak/cool and dwell: store up to 10 programs with 10 easy steps
- Adjustable over temperature shut-off for protocols that require setting of over temperature for protection – (class 2 according to norm DIN12880)
- Safe viewing of samples through internal glass door with limited impact on temperature
- Safe containment with automatic over and under temperature alarm
- Standard access port for data monitoring (large diameter of 42mm / 1.65 in.)
- Flexible shelf system for optimal loading, easy to remove for cleaning, with non-tip protection
- Stainless steel interior (1.4301 / ASTM 304) and rounded corners for easy cleaning
Specifications
| 13.5 cu. ft. | |
| Floor Model | |
| Stainless Steel (304/1.4301) | |
| Coated Steel | |
| N. America: Nema 5-15 plug w/ 120V 60Hz config.; All others: CEE7/7 with 230V 50Hz config. | |
| Automated Incubator | |
| Solid outer door & internal glass door | |
| CFC free/HFC free polyurethane insulation foam | |
| 1 | |
| Peltier technology: no refrigerant | |
| ≤0.3°C at 20°C; ≤0.7°C at 37°C | |
| 770 x 778 x 1545 mm (30.3 x 230.6 x 60.8 in.) | |
| 1545 mm (casters not included: 180 mm) | |
| 216 kg | |
| 167 kg | |
| 100/240 V |
| CE, CSA | |
| 381 L | |
| 20.79 x 21.06 in. (528 x 498 mm) | |
| 2 std./17 max. | |
| 5°C to 70°C | |
| Large Vacuum Fluorescent | |
| Energy efficient | |
| IMP400 | |
| 1.6 in. (42mm) | |
| 0.1°C | |
| 524 x 544 x 1335 mm (20.6 x 21.4 x 52.6 in.) | |
| 60.8 in. (casters not included: 7.1 in.) | |
| 476 lb. | |
| 368 lb. | |
| 100/240 V, 50/60 Hz, 3.8-9.0 A, 900 w |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.